2-Brom-3-hydroxypyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR6410-100G
OR6410-5G
OR6410-10G
APOSOR6410-100GEA
0
EUR
APOSOR6410-100G
APOSOR6410-5G
APOSOR6410-10G
2-Brom-3-hydroxypyridin 98%
2-Brom-3-hydroxypyridin
Formel:
C₅H₄BrNO Schmelzpunkt: 182…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006220 CAS-Nummer: 6602-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...