2,3-Dimethoxypyridin 96%

Lieferant: Apollo Scientific
Achtung

OR6412-10G
APOSOR6412-10GEA 90.2 EUR
APOSOR6412-10G
2,3-Dimethoxypyridin 96%
2,3-Dimethoxypyridine
Formel: C₇H₉NO₂
Molecular Weight: 139,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095043
CAS-Nummer: 52605-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR