1H-Imidazol-2-ylmethanol
Lieferant: Apollo Scientific
Synonyme:
2-(Hydroxymethyl)-1H-imidazole, (1H-Imidazol-2-yl)methanol
OR6415-1G
APOSOR6415-1GEA
91.5
EUR
APOSOR6415-1G
1H-Imidazol-2-ylmethanol
1H-Imidazol-2-ylmethanol
Formel:
C₄H₆N₂O Molecular Weight: 98.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202766 CAS-Nummer: 3724-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...