2,1,3-Benzoxadiazole-5-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Cyano-2,1,3-benzoxadiazole, Benzofurazan-5-carbonitrile

OR6422-1G
APOSOR6422-1GEA 143 EUR
APOSOR6422-1G
2,1,3-Benzoxadiazole-5-carbonitrile 98%
2,1,3-Benzoxadiazole-5-carbonitrile
Formel: C₇H₃N₃O
Molecular Weight: 145,118 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00206789
CAS-Nummer: 54286-62-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR