4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

OR6423-1G
APOSOR6423-1GEA 21.9 EUR
APOSOR6423-1G
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine
Formel: C₅H₃ClN₄
Molecular Weight: 154,5571 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03030404
CAS-Nummer: 5399-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR