1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole 95%

Lieferant: Apollo Scientific
Achtung

OR6435-250MG
APOSOR6435-250MGEA 91.5 EUR
APOSOR6435-250MG
1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole 95%
1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole

When melted, may contain small amount of sediment.

Formel: C₁₁H₁₀N₄O
Lagertemperatur: Kühlschrank
MDL: MFCD08235261
CAS-Nummer: 914637-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR