1-(Bromomethyl)-2,3-dimethylbenzene 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,3-Dimethylbenzyl bromide

OR6439-1G
APOSOR6439-1GEA 21.9 EUR
APOSOR6439-1G
1-(Bromomethyl)-2,3-dimethylbenzene 95%
1-(Bromomethyl)-2,3-dimethylbenzene
Formel: C₉H₁₁Br
Molecular Weight: 199,0876 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844741
CAS-Nummer: 81093-21-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR