2,5-Diaminopyridin Dihydrochlorid 96%

Lieferant: Apollo Scientific

OR6458-25G
APOSOR6458-25GEA 166 EUR
APOSOR6458-25G
2,5-Diaminopyridin Dihydrochlorid 96%
2,5-Diaminopyridin Dihydrochlorid
Formel: C₅H₉Cl₂N₃
Molecular Weight: 182,05 g/mol
Schmelzpunkt: 264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040818
CAS-Nummer: 26878-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR