3,4-Diaminopyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,4-DAP, 3,4-Pyridindiamin, Pyridin-3,4-diyldiamin
OR6459-100G
APOSOR6459-100GEA
296
EUR
APOSOR6459-100G
3,4-Diaminopyridin
3,4-Diaminopyridin
Formel:
C₅H₇N₃ Molecular Weight: 109,13 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006401 CAS-Nummer: 54-96-6 EINECS: 200-220-9 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...