2,2-Dibromopropanoic acid

Lieferant: Apollo Scientific

OR64743-500MG
APOSOR64743-500MGEA 292 EUR
APOSOR64743-500MG
2,2-Dibromopropanoic acid
2,2-Dibromopropanoic acid
Formel: C₃H₄Br₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD22053378
CAS-Nummer: 594-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR