3-(2,4,5-Trichlorophenyl)-1H-pyrazole

Lieferant: Apollo Scientific
Gefahr

OR64746-100MG OR64746-250MG
APOSOR64746-100MGEA 109 EUR
APOSOR64746-100MG APOSOR64746-250MG
3-(2,4,5-Trichlorophenyl)-1H-pyrazole
3-(2,4,5-Trichlorophenyl)-1H-pyrazole
Formel: C₉H₅Cl₃N₂
Molecular Weight: 247.51 g/mol
MDL: MFCD32862646
CAS-Nummer: 2645418-49-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR