3-(2-Iodophenyl)propanoic acid

Lieferant: Apollo Scientific

OR6479-5G
APOSOR6479-5GEA 219 EUR
APOSOR6479-5G
3-(2-Iodophenyl)propanoic acid
3-(2-Iodophenyl)propanoic acid

Radiopharmaceutical precursor

Formel: C₉H₉IO₂
Molecular Weight: 276.071 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040789
CAS-Nummer: 96606-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR