Phthalimidoacetaldehyddiethylacetal 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,2-Diethoxyethyl)isoindoline-1,3-dione, Phthalimidoacetaldehyd Diethylacetal
OR6499-25G
APOSOR6499-25GEA
63.8
EUR
APOSOR6499-25G
Phthalimidoacetaldehyddiethylacetal 98%
Phthalimidoacetaldehyddiethylacetal
Formel:
C₁₄H₁₇NO₄ Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005901 CAS-Nummer: 78902-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...