9-Hydroxyfluoren 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
9-Fluorenol
OR6505-25G
APOSOR6505-25GEA
71.2
EUR
APOSOR6505-25G
9-Hydroxyfluoren 97%
9-Hydroxyfluoren
Formel:
C₁₃H₁₀O Molecular Weight: 182,22 g/mol Schmelzpunkt: 155…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001135 CAS-Nummer: 1689-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...