4-Chloromethyl-5-methyl-2-(furan-2-yl)oxazole 95%

Lieferant: Apollo Scientific

OR6506-1G
APOSOR6506-1GEA 379 EUR
APOSOR6506-1G
4-Chloromethyl-5-methyl-2-(furan-2-yl)oxazole 95%
4-Chloromethyl-5-methyl-2-(furan-2-yl)oxazole
Formel: C₉H₈ClNO₂
Molecular Weight: 197,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056337
CAS-Nummer: 141399-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR