2-Aminoacetophenon Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR6519-100G OR6519-10G OR6519-25G
APOSOR6519-100GEA 256 EUR
APOSOR6519-100G APOSOR6519-10G APOSOR6519-25G
2-Aminoacetophenon Hydrochlorid
2-Aminoacetophenon Hydrochlorid
Formel: C₈H₁₀ClNO
Schmelzpunkt: 190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012873
CAS-Nummer: 5468-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR