2-Phthalimidopropionic acid 98%

Lieferant: Apollo Scientific

OR6527-25G
APOSOR6527-25GEA 106 EUR
APOSOR6527-25G
2-Phthalimidopropionic acid 98%
2-Phthalimidopropionic acid
Formel: C₁₁H₉NO₄
Schmelzpunkt: 159…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023061
CAS-Nummer: 19506-87-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR