1-Piperidinpropionsäure-Ethylester

Lieferant: Apollo Scientific

Synonyme: 3-(Piperidino)propionic Acid Ethyl Ester, Ethyl 3-(Piperidino)propionate, 1-Piperidinepropionic Acid Ethyl Ester

OR6540-100G OR6540-50G
APOSOR6540-100GEA 161 EUR
APOSOR6540-100G APOSOR6540-50G
1-Piperidinpropionsäure-Ethylester
Ethyl-1-piperidinepropionate
Formel: C₁₀H₁₉NO₂
Siedepunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006514
CAS-Nummer: 19653-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR