3-(1H-Benzimidazol-2-yl)-1-propanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6551-10G
OR6551-25G
OR6551-50G
OR6551-5G
OR6551-250MG
OR6551-500MG
APOSOR6551-10GEA
379
EUR
APOSOR6551-10G
APOSOR6551-25G
APOSOR6551-50G
APOSOR6551-5G
APOSOR6551-250MG
APOSOR6551-500MG
3-(1H-Benzimidazol-2-yl)-1-propanol 97%
2-(3-Hydroxypropyl)benzimidazole
Formel:
C₁₀H₁₂N₂O Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022688 CAS-Nummer: 2403-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...