3-(1H-Benzimidazol-2-yl)-1-propanol 97%

Lieferant: Apollo Scientific
Achtung

OR6551-10G OR6551-25G OR6551-50G OR6551-5G OR6551-250MG OR6551-500MG
APOSOR6551-10GEA 379 EUR
APOSOR6551-10G APOSOR6551-25G APOSOR6551-50G APOSOR6551-5G APOSOR6551-250MG APOSOR6551-500MG
3-(1H-Benzimidazol-2-yl)-1-propanol 97%
2-(3-Hydroxypropyl)benzimidazole
Formel: C₁₀H₁₂N₂O
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022688
CAS-Nummer: 2403-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR