trans-3-Chlorzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR6577-100G
OR6577-1KG
OR6577-500G
APOSOR6577-100GEA
106
EUR
APOSOR6577-100G
APOSOR6577-1KG
APOSOR6577-500G
trans-3-Chlorzimtsäure 98%
trans-3-Chlorzimtsäure
Formel:
C₉H₇ClO₂ Schmelzpunkt: 161…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004384 CAS-Nummer: 14473-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...