4-Chlorzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlorcinnamsäure, 3-(p-Chlorphenyl)propensäure, 3-(4-Chlorphenyl)propensäure

OR6578-25G
APOSOR6578-25GEA 21.9 EUR
APOSOR6578-25G
4-Chlorzimtsäure 98%
4-Chlorzimtsäure
Formel: C₉H₇ClO₂
Molecular Weight: 182,61 g/mol
Schmelzpunkt: 248…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004396
CAS-Nummer: 1615-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR