3-Pyridin-2-ylacrylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6591-500MG
APOSOR6591-500MGEA 14.7 EUR
APOSOR6591-500MG
3-Pyridin-2-ylacrylic acid 97%
3-Pyridin-2-ylacrylic acid
Formel: C₈H₇NO₂
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179422
CAS-Nummer: 7340-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR