Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
OR6605-100G
OR6605-25G
OR6605-5G
APOSOR6605-100GEA
65.4
EUR
APOSOR6605-100G
APOSOR6605-25G
APOSOR6605-5G
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat 98%
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat
Formel:
C₈H₉ClN₂O₂S Schmelzpunkt: 60…63 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006085 CAS-Nummer: 5909-24-0 EINECS: 227-619-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...