2,3-Diaminobenzoesäure

Lieferant: Apollo Scientific
Achtung

OR6615-25G
APOSOR6615-25GEA 280 EUR
APOSOR6615-25G
2,3-Diaminobenzoesäure
2,3-Diaminobenzoic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 178…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00137818
CAS-Nummer: 603-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR