tert-Butyl-2,6-dichloroisonicotinate 97%

Lieferant: Apollo Scientific

OR6618-5G
APOSOR6618-5GEA 63.8 EUR
APOSOR6618-5G
tert-Butyl-2,6-dichloroisonicotinate 97%
tert-Butyl-2,6-dichloroisonicotinate
Formel: C₁₀H₁₁Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08445655
CAS-Nummer: 75308-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR