4-Iod-1-methyl-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
OR6685-25G
OR6685-5G
OR6685-100G
APOSOR6685-25GEA
29
EUR
APOSOR6685-25G
APOSOR6685-5G
APOSOR6685-100G
4-Iod-1-methyl-1H-pyrazol
4-Iodo-1-methyl-1H-pyrazole
Formel:
C₄H₅IN₂ Schmelzpunkt: 59…64 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05663860 CAS-Nummer: 39806-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...