4-Dimethylamino-2-hydroxybenzaldehyde 98%

Lieferant: Apollo Scientific

OR6690-1G OR6690-5G
APOSOR6690-1GEA 60.1 EUR
APOSOR6690-1G APOSOR6690-5G
4-Dimethylamino-2-hydroxybenzaldehyde 98%
4-Dimethylamino-2-hydroxybenzaldehyde
Formel: C₉H₁₁NO₂
Schmelzpunkt: 78 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00027419
CAS-Nummer: 41602-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR