4-Dimethylamino-2-hydroxybenzaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Dimethylamino)-2-hydroxybenzaldehyde, 4-(Dimethylamino)salicylaldehyde, 5-(Dimethylamino)-2-formylphenol

OR6690-5G
APOSOR6690-5GEA 142 EUR
APOSOR6690-5G
4-Dimethylamino-2-hydroxybenzaldehyde 98%
4-Dimethylamino-2-hydroxybenzaldehyde
Formel: C₉H₁₁NO₂
Molecular Weight: 165.189 g/mol
Schmelzpunkt: 78 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00027419
CAS-Nummer: 41602-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR