1-Brom-2-(brommethyl)naphthalin
Lieferant: Apollo Scientific
|
Gefahr
|
OR6694-100MG
APOSOR6694-100MGEA
23.2
EUR
APOSOR6694-100MG
1-Brom-2-(brommethyl)naphthalin
1-Bromo-2-(bromomethyl)naphthalene
Formel:
C₁₁H₈Br₂ Molecular Weight: 299,99 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046369 CAS-Nummer: 37763-43-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...