3-Hydroxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
m-Hydroxybenzonitril, 3-Cyanphenol, m-Cyanphenol
OR6704-25G
OR6704-5G
OR6704-1G
APOSOR6704-25GEA
45.1
EUR
APOSOR6704-25G
APOSOR6704-5G
APOSOR6704-1G
3-Hydroxybenzonitril
3-Hydroxybenzonitril
Formel:
C₇H₅NO Molecular Weight: 119.12 g/mol Siedepunkt: 265 Schmelzpunkt: 78…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002252 CAS-Nummer: 873-62-1 EINECS: 212-845-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...