5,6-Dimethylbenzimidazol
Lieferant: Apollo Scientific
|
Achtung
|
OR6715-100G
OR6715-500G
APOSOR6715-100GEA
56.7
EUR
APOSOR6715-100G
APOSOR6715-500G
5,6-Dimethylbenzimidazol
5,6-Dimethylbenzimidazole
Formel:
C₉H₁₀N₂ Schmelzpunkt: 202…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005603 CAS-Nummer: 582-60-5 EINECS: 209-488-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...