4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Methyl-1-phenyl-1H-pyrazol-4-yl isocyanate, (4-Isocyanato-5-methyl-1H-pyrazol-4-yl)benzene

OR6719-250MG
APOSOR6719-250MGEA 140 EUR
APOSOR6719-250MG
4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole 97%
4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole

May form small amount of sediment

Formel: C₁₁H₉N₃O
Molecular Weight: 199,209 g/mol
Schmelzpunkt: 56…60 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681922
CAS-Nummer: 799283-97-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR