5-Methyl-3-phenylisoxazole-4-sulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR6722-1G
APOSOR6722-1GEA
0
EUR
APOSOR6722-1G
5-Methyl-3-phenylisoxazole-4-sulphonyl chloride 97%
5-Methyl-3-phenylisoxazole-4-sulphonyl chloride
Formel:
C₁₀H₈ClNO₃S Schmelzpunkt: 61…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658966 CAS-Nummer: 857283-56-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...