1,3-benzodioxole-5-sulphonyl chloride 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR6723-1G
OR6723-250MG
APOSOR6723-1GEA
81.2
EUR
APOSOR6723-1G
APOSOR6723-250MG
1,3-benzodioxole-5-sulphonyl chloride 95+%
1,3-benzodioxole-5-sulphonyl chloride
Formel:
C₇H₅ClO₄S Schmelzpunkt: 48…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974524 CAS-Nummer: 115010-10-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...