N-[4-(1H-Imidazol-1-yl)benzyl]-N-methylamine 95%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-4-(1-imidazolyl)benzylamine

OR6725-1G
APOSOR6725-1GEA 201 EUR
APOSOR6725-1G
N-[4-(1H-Imidazol-1-yl)benzyl]-N-methylamine 95%
N-[4-(1H-Imidazol-1-yl)benzyl]-N-methylamine
Formel: C₁₁H₁₃N₃
Molecular Weight: 187,241 g/mol
Schmelzpunkt: 67…69,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772795
CAS-Nummer: 179873-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR