[2-(1H-Pyrazol-1-yl)phenyl]methylamine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-[2-(Aminomethyl)phenyl]-1H-pyrazole, 1-(Aminomethyl)-2-(1H-pyrazol-1-yl)benzene, 2-(1H-Pyrazol-1-yl)benzylamine
OR6729-1G
APOSOR6729-1GEA
194
EUR
APOSOR6729-1G
[2-(1H-Pyrazol-1-yl)phenyl]methylamine 95%
[2-(1H-Pyrazol-1-yl)phenyl]methylamine
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,214 g/mol Siedepunkt: 100 °C (0,5 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086189 CAS-Nummer: 449758-13-8 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...