1-Methyl-2-formylbenzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-benzo[d]imidazole-2-carbaldehyde

OR6737-1G
APOSOR6737-1GEA 116 EUR
APOSOR6737-1G
1-Methyl-2-formylbenzimidazole
1-Methyl-2-formylbenzimidazole
Formel: C₉H₈N₂O
Molecular Weight: 160.176 g/mol
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142655
CAS-Nummer: 3012-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR