Thieno[2,3-b]pyridine-2-carboxaldehyde 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Thieno[2,3-b]pyridine-2-carbaldehyde, 2-Formylthieno[2,3-b]pyridine

OR6738-100MG
APOSOR6738-100MGEA 0 EUR
APOSOR6738-100MG
Thieno[2,3-b]pyridine-2-carboxaldehyde 97+%
Thieno[2,3-b]pyridine-2-carboxaldehyde
Formel: C₈H₅NOS
Molecular Weight: 163.196 g/mol
Schmelzpunkt: 141.5…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772791
CAS-Nummer: 53174-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR