2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,5-Dimethyl-1,3-oxazol-4-yl)methanol, (2,5-Dimethyloxazol-4-yl)methanol

OR6751-1G OR6751-250MG OR6751-5G
APOSOR6751-1GEA 139 EUR
APOSOR6751-1G APOSOR6751-250MG APOSOR6751-5G
2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole 97%
2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole
Formel: C₆H₉NO₂
Molecular Weight: 127.141 g/mol
Schmelzpunkt: 73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772804
CAS-Nummer: 92901-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR