1-(8-Chloroisoquinolin-5-ylsulphonyl)piperazine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: HA-156

OR6751T-10MG
APOSOR6751T-10MGEA 288 EUR
APOSOR6751T-10MG
1-(8-Chloroisoquinolin-5-ylsulphonyl)piperazine dihydrochloride
1-(8-Chloroisoquinolin-5-ylsulphonyl)piperazine dihydrochloride

Inhinbits calcium-ion dependent myosin phosphorylation by MLC-Kinase & protein kinase C.

Formel: C₁₃H₁₄ClN₃O₂S·2ClH
Molecular Weight: 384,709 g/mol
Schmelzpunkt: >240 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00269948
CAS-Nummer: 355115-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR