4-[(Piperidin-1-yl)methyl]benzyl alcohol

Lieferant: Apollo Scientific
Gefahr

Synonyme: [4-(Piperidin-1-ylmethyl)phenyl]methanol

OR6752-1G
APOSOR6752-1GEA 116 EUR
APOSOR6752-1G
4-[(Piperidin-1-yl)methyl]benzyl alcohol
4-[(Piperidin-1-yl)methyl]benzyl alcohol
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772863
CAS-Nummer: 91271-62-4
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR