Methyl 4-amino-3,5-dichlorobenzoate 97%
Lieferant: Apollo Scientific
OR6766-5G
APOSOR6766-5GEA
101
EUR
APOSOR6766-5G
Methyl 4-amino-3,5-dichlorobenzoate 97%
Methyl 4-amino-3,5-dichlorobenzoate
Formel:
C₈H₇Cl₂NO₂ Molecular Weight: 220,053 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436086 CAS-Nummer: 41727-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...