4-Biphenylmethyl chloride

Lieferant: Apollo Scientific
Achtung

OR6777-1G OR6777-25G OR6777-5G
APOSOR6777-1GEA 0 EUR
APOSOR6777-1G APOSOR6777-25G APOSOR6777-5G
4-Biphenylmethyl chloride
4-Biphenylmethyl chloride
Formel: C₁₃H₁₁Cl
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000917
CAS-Nummer: 1667-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR