5-Brom-1,2,3-trimethoxybenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-3,4,5-trimethoxybenzol

OR6783-100G
APOSOR6783-100GEA 208 EUR
APOSOR6783-100G
5-Brom-1,2,3-trimethoxybenzol 98%
5-Brom-1,2,3-trimethoxybenzol
Formel: C₉H₁₁BrO₃
Molecular Weight: 247.09 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017169
CAS-Nummer: 2675-79-8
EINECS: 220-223-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR