[3-(Piperidin-1-ylmethyl)phenyl]amine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-[(Piperidin-1-yl)methyl]aniline

OR6791-1G
APOSOR6791-1GEA 78.6 EUR
APOSOR6791-1G
[3-(Piperidin-1-ylmethyl)phenyl]amine 97%
[3-(Piperidin-1-ylmethyl)phenyl]amine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Schmelzpunkt: 111…113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04114502
CAS-Nummer: 93138-55-7
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR