3-(Morpholinomethyl)aniline
Lieferant: Apollo Scientific
OR6794-1G
APOSOR6794-1GEA
58
EUR
APOSOR6794-1G
3-(Morpholinomethyl)aniline
3-(Morpholinomethyl)aniline
Formel:
C11H16N2O Molecular Weight: 192.26 g/mol Schmelzpunkt: 137…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03074538 CAS-Nummer: 123207-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...