N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine 97%

Lieferant: Apollo Scientific

OR6797-1G
APOSOR6797-1GEA 322 EUR
APOSOR6797-1G
N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine 97%
N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine
Formel: C₁₄H₂₂N₂O₂
Molecular Weight: 250,337 g/mol
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060519
CAS-Nummer: 857284-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR