2-(Thien-2-yl)benzylamine hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[2-(Thien-2-yl)phenyl]methylamine hydrochloride, 2-[2-(Aminomethyl)phenyl]thiophene hydrochloride
OR6806-1G
APOSOR6806-1GEA
205
EUR
APOSOR6806-1G
2-(Thien-2-yl)benzylamine hydrochloride 95%
2-(Thien-2-yl)benzylamine hydrochloride
Formel:
C₁₁H₁₁NS·ClH Schmelzpunkt: 186,5…188.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07772821 CAS-Nummer: 863991-95-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...