1-(1,3-Thiazol-2-yl)thiourea

Lieferant: Apollo Scientific
Achtung

OR6809-1G
APOSOR6809-1GEA 574 EUR
APOSOR6809-1G
1-(1,3-Thiazol-2-yl)thiourea
1-(1,3-Thiazol-2-yl)thiourea
Formel: C₄H₅N₃S₂
Lagertemperatur: Kühlschrank
MDL: MFCD04971712
CAS-Nummer: 19958-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR